siRNA Details
| Gene Symbol: C4bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007576.3 | NP_031602.3 | 6, 7 | 923 | ||
| GenBank mRNA | AB094485.1 | - | 904 | |||
| AB094486.1 | - | 6, 7 | 839 | |||
| AK136735.1 | - | 6, 7 | 837 | |||
| AK149468.1 | - | 6, 7 | 930 | |||
| AK149477.1 | - | 6, 7 | 840 | |||
| AK162345.1 | - | 6, 7 | 837 | |||
| BC012257.1 | - | 6, 7 | 895 | 4163203 | ||
| M17122.1 | - | 6, 7 | 937 | |||

C4bp
Genomic Map