siRNA Details
| Gene Symbol: C9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013485.1 | NP_038513.1 | 8 | 1260 | ||
| GenBank mRNA | BC011137.1 | - | 8 | 1260 | 4195787 | |
| X05475.1 | - | 8 | 1239 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9| Gene Symbol: C9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013485.1 | NP_038513.1 | 8 | 1260 | ||
| GenBank mRNA | BC011137.1 | - | 8 | 1260 | 4195787 | |
| X05475.1 | - | 8 | 1239 | |||