siRNA Details
Gene Symbol: C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013485.1 | NP_038513.1 | 5 | 645 | ||
GenBank mRNA | BC011137.1 | - | 5 | 645 | 4195787 | |
X05475.1 | - | 5 | 624 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013485.1 | NP_038513.1 | 5 | 645 | ||
GenBank mRNA | BC011137.1 | - | 5 | 645 | 4195787 | |
X05475.1 | - | 5 | 624 |