siRNA Details
Gene Symbol: Cacna1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB086123.1 | - | 35 | 4214 | ||
AJ437291.1 | - | 33 | 4091 | |||
AJ437292.1 | - | 33 | 4212 | |||
AK147415.1 | - | 34 | 5682 | |||
BC058783.1 | - | 3 | 440 | |||
M57975.1 | - | 7 | 650 |
