siRNA Details
Gene Symbol: Cacna1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB086123.1 | - | 40, 41 | 4868 | ||
AJ437291.1 | - | 38, 39 | 4745 | |||
AJ437292.1 | - | 38, 39 | 4866 | |||
AK147415.1 | - | 39, 40 | 6336 | |||
BC058783.1 | - | 8, 9 | 1094 |
