siRNA Details
| Gene Symbol: Cacna1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009782.3 | NP_033912.2 | 11 | 1456 | ||
| GenBank mRNA | BC150750.1 | - | 11 | 1567 | 9087661 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacna1e| Gene Symbol: Cacna1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009782.3 | NP_033912.2 | 11 | 1456 | ||
| GenBank mRNA | BC150750.1 | - | 11 | 1567 | 9087661 | |