siRNA Details
Gene Symbol: Cacna1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009782.3 | NP_033912.2 | 37 | 5103 | ||
GenBank mRNA | BC150750.1 | - | 37 | 5214 | 9087661 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cacna1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009782.3 | NP_033912.2 | 37 | 5103 | ||
GenBank mRNA | BC150750.1 | - | 37 | 5214 | 9087661 |