siRNA Details
Gene Symbol: Cacna2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009785.1 | NP_033915.1 | 11 | 1227 | ||
GenBank mRNA | AJ010949.1 | - | 11 | 1227 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cacna2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009785.1 | NP_033915.1 | 11 | 1227 | ||
GenBank mRNA | AJ010949.1 | - | 11 | 1227 |