siRNA Details
| Gene Symbol: Cacnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252533.1 | NP_001239462.1 | 12 | 1446 | ||
| NM_023116.4 | NP_075605.1 | 13 | 1577 | |||
| GenBank mRNA | AK034054.1 | - | 13 | 1578 | ||
| AM259383.2 | - | 12 | 1446 | |||
| BC109156.1 | - | 13 | 1509 | 40047524 | ||
| BC109157.1 | - | 13 | 1509 | 40047531 | ||
| CF173155.1 | - | 2 | 556 | |||
| KJ789960.1 | - | 12 | 1165 | |||
| L20343.1 | - | 12 | 1356 | |||

Cacnb2
Genomic Map