siRNA Details
| Gene Symbol: Camk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009793.3 | NP_033923.2 | 4 | 685 | ||
| GenBank mRNA | AK030219.1 | - | 4 | 569 | ||
| AK043114.1 | - | 4 | 395 | |||
| BC070420.1 | - | 4 | 566 | 5718423 | ||
| X58995.1 | - | 4 | 375 | |||

Camk4
Genomic Map