siRNA Details
| Gene Symbol: Camk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009793.3 | NP_033923.2 | 11 | 1751 | ||
| GenBank mRNA | AK030219.1 | - | 11 | 1635 | ||
| AK040523.1 | - | 1 | 46 | |||
| AK043114.1 | - | 11 | 1461 | |||
| BC070420.1 | - | 11 | 1632 | 5718423 | ||
| J03057.1 | - | 3 | 654 | |||
| M16206.1 | - | 2 | 454 | |||
| M64266.1 | - | 3 | 702 | |||
| X58995.1 | - | 11 | 1441 | |||

Camk4
Genomic Map