siRNA Details
| Gene Symbol: Capn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007603.3 | NP_031629.3 | 6 | 1053 | ||
| GenBank mRNA | AK145116.1 | - | 6 | 1068 | ||
| BC052167.1 | - | 6 | 954 | 6517565 | ||
| Y12582.1 | - | 6 | 962 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capn6| Gene Symbol: Capn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007603.3 | NP_031629.3 | 6 | 1053 | ||
| GenBank mRNA | AK145116.1 | - | 6 | 1068 | ||
| BC052167.1 | - | 6 | 954 | 6517565 | ||
| Y12582.1 | - | 6 | 962 | |||