siRNA Details
| Gene Symbol: Casp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042072.1 | - | 6 | 666 | ||
| AK133829.1 | - | 6 | 677 | |||
| AK170649.1 | - | 5 | 399 | |||
| BC034262.1 | - | 6 | 732 | 4460089 | ||
| D28492.1 | - | 6 | 655 | |||
| Y13085.1 | - | 6 | 734 | |||

Casp2
Genomic Map