siRNA Details
| Gene Symbol: Casp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277932.1 | NP_001264861.1 | 8 | 1442 | ||
| NM_015733.5 | NP_056548.2 | 9 | 1552 | |||
| GenBank mRNA | AB019600.1 | - | 9 | 1309 | ||
| AB019601.1 | - | 8 | 1199 | |||
| AK085095.1 | - | 9 | 1341 | |||
| AK085152.1 | - | 9 | 1308 | |||
| AK154495.1 | - | 9 | 1322 | |||
| BC056372.1 | - | 9 | 1334 | 6834067 | ||
| BC056447.1 | - | 9 | 1552 | 5721188 | ||
| CT010400.1 | - | 9 | 1309 | |||

Casp9
Genomic Map