siRNA Details
| Gene Symbol: Casr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013803.2 | NP_038831.2 | 7 | 2793 | ||
| GenBank mRNA | AB027140.1 | - | 2 | 588 | ||
| AF110178.1 | - | 7 | 2794 | |||
| AF110179.1 | - | 6 | 2563 | |||
| AF128842.1 | - | 6 | 2491 | |||
| AK029770.1 | - | 4 | 1382 | |||
More Information
Set Membership: |
|

Casr
Genomic Map