siRNA Details
| Gene Symbol: Runx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146038.2 | NP_001139510.1 | 1 | 86 | ||
| NM_001271627.1 | NP_001258556.1 | 1 | 86 | |||
| NM_009820.5 | NP_033950.2 | 1 | 86 | |||
| GenBank mRNA | AF005936.1 | - | 1 | 40 | ||
| AF010284.1 | - | 1 | 86 | |||
| AF020681.1 | - | 1 | 86 | |||
| AF053951.1 | - | 1 | 86 | |||
| AF053954.1 | - | 1 | 86 | |||
| DQ458792.1 | - | 1 | 257 | |||

Runx2
Genomic Map