siRNA Details
| Gene Symbol: Runx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145920.2 | NP_001139392.1 | 7 | 2530 | ||
| NM_001146038.2 | NP_001139510.1 | 9 | 1795 | |||
| NM_001271627.1 | NP_001258556.1 | 8 | 1959 | |||
| NM_001271630.1 | NP_001258559.1 | 6 | 1884 | |||
| NM_001271631.1 | NP_001258560.1 | 5 | 1779 | |||
| NM_009820.5 | NP_033950.2 | 9 | 1762 | |||
| GenBank mRNA | AF005936.1 | - | 8 | 1913 | ||
| AF010284.1 | - | 9 | 1762 | |||
| AK089291.1 | - | 5 | 1317 | |||
| D14636.1 | - | 7 | 2528 | |||

Runx2
Genomic Map