siRNA Details
| Gene Symbol: Cckar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009827.2 | NP_033957.1 | 2 | 486 | ||
| GenBank mRNA | AK004730.2 | - | 2 | 344 | ||
| AK143895.1 | - | 2 | 334 | |||
| AK146738.1 | - | 2 | 487 | |||
| AK147052.1 | - | 2 | 487 | |||
| AK164301.1 | - | 78 | ||||
| BC020534.1 | - | 2 | 306 | 4236240 | ||
More Information
Set Membership: |
|

Cckar
Genomic Map