siRNA Details
Gene Symbol: Cd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043317.2 | NP_001036782.1 | 10 | 2048 | ||
NM_009845.3 | NP_033975.3 | 8 | 1923 | |||
GenBank mRNA | AF102134.1 | - | 7 | 1692 | ||
AF115400.1 | - | 7 | 1692 | |||
AF115401.1 | - | 7 | 1692 | |||
AK041230.1 | - | 10 | 2055 | |||
AK143835.1 | - | 10 | 1986 | |||
AK156735.1 | - | 8 | 1922 | |||
AK171589.1 | - | 7 | 1729 | |||
AK172301.1 | - | 8 | 1879 | |||
BC046957.1 | - | 10 | 1993 | |||
BC150807.1 | - | 9 | 2007 | 9087718 | ||
L02844.1 | - | 9 | 2194 | |||
L16928.1 | - | 9 | 2171 |
