siRNA Details
| Gene Symbol: Scarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007644.3 | NP_031670.1 | 8 | 1311 | ||
| GenBank mRNA | AB008553.1 | - | 8 | 1216 | ||
| AK011123.1 | - | 8 | 1241 | |||
| AK083038.1 | - | 8 | 1312 | |||
| AK144235.1 | - | 8 | 1343 | |||
| AK149102.1 | - | 1258 | ||||
| AK185315.1 | - | 2 | 154 | |||
| BC029073.1 | - | 8 | 1219 | 3962669 | ||

Scarb2
Genomic Map