siRNA Details
| Gene Symbol: Cd37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290802.1 | NP_001277731.1 | 8, 9 | 940 | ||
| NM_001290804.1 | NP_001277733.1 | 7, 8 | 865 | |||
| NM_007645.4 | NP_031671.1 | 7, 8 | 889 | |||
| GenBank mRNA | AK154466.1 | - | 8, 9 | 910 | ||
| AK155663.1 | - | 6 | 2211 | |||
| AK155966.1 | - | 7, 8 | 854 | |||
| BC019402.1 | - | 7, 8 | 926 | 4013070 | ||
| BY548149.1 | - | 1, 2 | 101 | |||
| CT010217.1 | - | 7, 8 | 778 | |||

Cd37
Genomic Map