siRNA Details
| Gene Symbol: Cd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013488.2 | NP_038516.1 | 4 | 448 | ||
| GenBank mRNA | AF045882.1 | - | 2 | 207 | ||
| AK040115.1 | - | 4 | 440 | |||
| AK153966.1 | - | 4 | 448 | |||
| AK161775.1 | - | 4 | 372 | |||
| BC039137.1 | - | 4 | 427 | 3670597 | ||
| M13816.1 | - | 3 | 318 | |||
| M36850.1 | - | 3 | 318 | |||
| X04836.1 | - | 4 | 448 | |||

Cd4
Genomic Map