siRNA Details
| Gene Symbol: Cd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009854.2 | NP_033984.1 | 2 | 359 | ||
| GenBank mRNA | AK136444.1 | - | 2 | 313 | ||
| AK154339.1 | - | 2 | 314 | |||
| BC024376.1 | - | 2 | 300 | 4985794 | ||
| CT010368.1 | - | 2 | 246 | |||
| D10329.1 | - | 2 | 299 | |||

Cd7
Genomic Map