siRNA Details
| Gene Symbol: Cd72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110320.1 | NP_001103790.1 | 7 | 991 | ||
| NM_001110321.1 | NP_001103791.1 | 6 | 919 | |||
| NM_001110322.1 | NP_001103792.1 | 5 | 808 | |||
| NM_007654.2 | NP_031680.2 | 7 | 970 | |||
| GenBank mRNA | AA203771.1 | - | 2 | 144 | ||
| AF543214.1 | - | 5 | 667 | |||
| AK134132.1 | - | 7 | 994 | |||
| AK144191.1 | - | 6 | 922 | |||
| AK151094.1 | - | 810 | ||||
| BC003824.1 | - | 7 | 928 | 3599554 | ||
| J04170.1 | - | 7 | 907 | |||
| S40777.1 | - | 7 | 924 | |||

Cd72
Genomic Map