siRNA Details
| Gene Symbol: Cd81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133655.2 | NP_598416.1 | 3, 4 | 539 | ||
| GenBank mRNA | AK136370.1 | - | 3, 4 | 503 | ||
| AK166521.1 | - | 3, 4 | 505 | |||
| AK170741.1 | - | 3, 4 | 504 | |||
| AK182212.1 | - | 11 | ||||
| AK185682.1 | - | 11 | ||||
| AK211761.1 | - | 11 | ||||
| BC011433.1 | - | 3, 4 | 494 | 2811801 | ||
| X59047.1 | - | 3, 4 | 259 | |||

Cd81
Genomic Map