siRNA Details
| Gene Symbol: Cd81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133655.2 | NP_598416.1 | 8 | 911 | ||
| GenBank mRNA | AK136370.1 | - | 8 | 875 | ||
| AK166521.1 | - | 8 | 877 | |||
| AK170741.1 | - | 8 | 876 | |||
| AK183391.1 | - | 3 | 127 | |||
| AK195226.1 | - | 127 | ||||
| BC011433.1 | - | 8 | 866 | 2811801 | ||
| BQ175686.1 | - | 1 | 620 | |||
| BQ175800.1 | - | 1 | 620 | |||
| X59047.1 | - | 8 | 631 | |||

Cd81
Genomic Map