siRNA Details
| Gene Symbol: Cd86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019388.3 | NP_062261.3 | 4 | 661 | ||
| GenBank mRNA | AF065897.1 | - | 4 | 577 | ||
| AF065898.1 | - | 4 | 577 | |||
| AF065899.1 | - | 4 | 577 | |||
| AF065900.1 | - | 4 | 577 | |||
| AK079513.1 | - | 4 | 662 | |||
| AK170054.1 | - | 4 | 662 | |||
| AK172548.1 | - | 4 | 664 | |||
| BC013807.1 | - | 4 | 635 | 4008635 | ||
| L25606.1 | - | 4 | 643 | |||
| S70108.1 | - | 4 | 626 | |||

Cd86
Genomic Map