siRNA Details
| Gene Symbol: Cdc25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007658.3 | NP_031684.3 | 3 | 683 | ||
| GenBank mRNA | AK077609.1 | - | 3 | 300 | ||
| AK077991.1 | - | 3 | 686 | |||
| AK142334.1 | - | 2 | 958 | |||
| BC046296.1 | - | 3 | 302 | 6486343 | ||
| U27323.1 | - | 3 | 269 | |||

Cdc25a
Genomic Map