siRNA Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111075.4 | NP_001104545.1 | 12 | 1480 | ||
| NM_023117.4 | NP_075606.1 | 13 | 1558 | |||
| GenBank mRNA | AK004836.1 | - | 12 | 1437 | ||
| AK043730.1 | - | 12 | 1471 | |||
| AK153903.1 | - | 13 | 1559 | |||
| AK157397.1 | - | 12 | 1438 | |||
| BC002287.1 | - | 4 | 345 | |||
| BC048164.1 | - | 11 | 1638 | |||
| BC057568.1 | - | 12 | 1351 | 6401489 | ||
| S93521.1 | - | 13 | 1427 | |||

Cdc25b
Genomic Map