siRNA Details
| Gene Symbol: Cdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007659.3 | NP_031685.2 | 4 | 329 | ||
| GenBank mRNA | AK030231.1 | - | 4 | 330 | ||
| AK051859.1 | - | 4 | 356 | |||
| AK135516.1 | - | 4 | 333 | |||
| AK168054.1 | - | 4 | 330 | |||
| BC005614.1 | - | 3 | 222 | |||
| BC024396.1 | - | 4 | 307 | 4977366 | ||
| M38724.1 | - | 3 | 250 | |||
| U58633.1 | - | 4 | 333 | |||
| X16461.1 | - | 3 | 250 | |||

Cdk1
Genomic Map