siRNA Details
| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007661.3 | NP_031687.2 | 17 | 1969 | ||
| GenBank mRNA | AK077668.1 | - | 17 | 1970 | ||
| AK147133.1 | - | 17 | 1947 | |||
| BC025058.1 | - | 6 | 619 | |||
| BC052920.1 | - | 17 | 2047 | 6489832 | ||
| L37092.1 | - | 16 | 2541 | |||
| M58633.1 | - | 12 | 1312 | |||

Cdk11b
Genomic Map