siRNA Details
| Gene Symbol: Arhgap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020260.2 | NP_064656.2 | 12 | 4270 | ||
| GenBank mRNA | AK129309.1 | - | 3 | 2695 | ||
| AK166583.1 | - | 1 | 452 | |||
| AK166622.1 | - | 1 | 452 | |||
| BC138870.1 | - | 12 | 4075 | 8861892 | ||

Arhgap31
Genomic Map