siRNA Details
| Gene Symbol: Cdh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007666.3 | NP_031692.2 | 12 | 2336 | ||
| GenBank mRNA | BC107008.1 | - | 11 | 2077 | 40047565 | |
| BC107013.1 | - | 11 | 2077 | 40048308 | ||
| D82029.1 | - | 11 | 2077 | |||

Cdh6
Genomic Map