siRNA Details
| Gene Symbol: Cdh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007666.3 | NP_031692.2 | 8 | 1565 | ||
| GenBank mRNA | BC107008.1 | - | 7 | 1306 | 40047565 | |
| BC107013.1 | - | 7 | 1306 | 40048308 | ||
| D82029.1 | - | 7 | 1306 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh6| Gene Symbol: Cdh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007666.3 | NP_031692.2 | 8 | 1565 | ||
| GenBank mRNA | BC107008.1 | - | 7 | 1306 | 40047565 | |
| BC107013.1 | - | 7 | 1306 | 40048308 | ||
| D82029.1 | - | 7 | 1306 | |||