siRNA Details
| Gene Symbol: Cdyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123386.1 | NP_001116858.1 | 6, 7 | 1943 | ||
| NM_009881.3 | NP_034011.1 | 9, 10 | 1893 | |||
| GenBank mRNA | AF081260.1 | - | 9, 10 | 1875 | ||
| AF081261.1 | - | 9, 10 | 1875 | |||
| AK028162.1 | - | 6, 7 | 1639 | |||
| AK156509.1 | - | 6, 7 | 1944 | |||
| AK162180.1 | - | 4, 5 | 623 | |||
| BC055103.1 | - | 9, 10 | 1842 | 6741468 | ||
| BC062123.1 | - | 6, 7 | 2032 | 6826210 | ||

Cdyl
Genomic Map