siRNA Details
| Gene Symbol: Cenpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007683.3 | NP_031709.2 | 10 | 1856 | ||
| GenBank mRNA | BC150700.1 | - | 10 | 1652 | 9087611 | |
| U03113.1 | - | 10 | 1742 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpc1| Gene Symbol: Cenpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007683.3 | NP_031709.2 | 10 | 1856 | ||
| GenBank mRNA | BC150700.1 | - | 10 | 1652 | 9087611 | |
| U03113.1 | - | 10 | 1742 | |||