siRNA Details
| Gene Symbol: Cfi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007686.2 | NP_031712.2 | 12 | 1347 | ||
| GenBank mRNA | AA530545.1 | - | 3 | 237 | ||
| BC150751.1 | - | 12 | 1340 | 9087662 | ||
| U47810.1 | - | 12 | 1350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfi| Gene Symbol: Cfi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007686.2 | NP_031712.2 | 12 | 1347 | ||
| GenBank mRNA | AA530545.1 | - | 3 | 237 | ||
| BC150751.1 | - | 12 | 1340 | 9087662 | ||
| U47810.1 | - | 12 | 1350 | |||