siRNA Details
| Gene Symbol: Cfi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007686.2 | NP_031712.2 | 13, 14 | 1674 | ||
| GenBank mRNA | BC150751.1 | - | 13, 14 | 1667 | 9087662 | |
| BG072327.1 | - | 1, 2 | 396 | |||
| U47810.1 | - | 13, 14 | 1677 | |||

Cfi
Genomic Map