siRNA Details
| Gene Symbol: Cfi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007686.2 | NP_031712.2 | 6 | 909 | ||
| GenBank mRNA | BC150751.1 | - | 6 | 902 | 9087662 | |
| CN691735.1 | - | 3 | 361 | |||
| U47810.1 | - | 6 | 912 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfi| Gene Symbol: Cfi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007686.2 | NP_031712.2 | 6 | 909 | ||
| GenBank mRNA | BC150751.1 | - | 6 | 902 | 9087662 | |
| CN691735.1 | - | 3 | 361 | |||
| U47810.1 | - | 6 | 912 | |||