siRNA Details
| Gene Symbol: Chek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007691.5 | NP_031717.2 | 9, 10 | 1026 | ||
| GenBank mRNA | AF016583.1 | - | 8, 9 | 935 | ||
| AF032875.1 | - | 3, 4 | 304 | |||
| AK011258.1 | - | 9, 10 | 970 | |||
| AK033179.1 | - | 9, 10 | 996 | |||
| AK045336.1 | - | 9, 10 | 968 | |||
| AK166675.1 | - | 9, 10 | 1027 | |||
| BC037613.1 | - | 8, 9 | 739 | |||
| BC053394.1 | - | 9, 10 | 963 | |||
| BC064445.1 | - | 8, 9 | 1425 | |||
| BC100386.1 | - | 8, 9 | 1101 | 30481390 | ||

Chek1
Genomic Map