siRNA Details
| Gene Symbol: Chek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007691.5 | NP_031717.2 | 11 | 1259 | ||
| GenBank mRNA | AF016583.1 | - | 10 | 1168 | ||
| AK011258.1 | - | 11 | 1203 | |||
| AK033179.1 | - | 11 | 1229 | |||
| AK045336.1 | - | 11 | 1201 | |||
| AK166675.1 | - | 11 | 1260 | |||
| BC037613.1 | - | 10 | 972 | |||
| BC053394.1 | - | 11 | 1196 | |||
| BC064445.1 | - | 10 | 1658 | |||
| BC100386.1 | - | 10 | 1334 | 30481390 | ||

Chek1
Genomic Map