siRNA Details
| Gene Symbol: Chil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007695.3 | NP_031721.2 | 5 | 509 | ||
| GenBank mRNA | AK051475.1 | - | 5 | 450 | ||
| AK155412.1 | - | 5 | 449 | |||
| BC003780.1 | - | 5 | 445 | 3601796 | ||
| BC004734.1 | - | 5 | 436 | 3583267 | ||
| BC005611.1 | - | 5 | 456 | 3582304 | ||
| CT010283.1 | - | 5 | 385 | |||
| EU313768.1 | - | 5 | 385 | |||
| X93035.1 | - | 5 | 450 | |||

Chil1
Genomic Map