siRNA Details
| Gene Symbol: Ckmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009897.2 | NP_034027.1 | 8 | 1312 | ||
| GenBank mRNA | AK005076.1 | - | 8 | 1311 | ||
| AK131953.1 | - | 8 | 1316 | |||
| BC025976.1 | - | 9 | 1227 | 4216758 | ||
| Z13968.1 | - | 8 | 1312 | |||

Ckmt1
Genomic Map