siRNA Details
| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013491.2 | NP_038519.1 | 2 | 357 | ||
| GenBank mRNA | AK036735.1 | - | 2 | 359 | ||
| AY046403.1 | - | 2 | 182 | |||
| AY046404.1 | - | 2 | 182 | |||
| BC114336.1 | - | 2 | 322 | 40048966 | ||

Clcn1
Genomic Map