siRNA Details
| Gene Symbol: Clcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009900.2 | NP_034030.2 | 10, 11 | 1748 | ||
| GenBank mRNA | AF097415.1 | - | 10, 11 | 1226 | ||
| AF139724.1 | - | 10, 11 | 1177 | |||
| AK141656.1 | - | 11, 12 | 2384 | |||
| AK155104.1 | - | 10, 11 | 1042 | |||
| AK165000.1 | - | 10, 11 | 1276 | |||
| AK171231.1 | - | 10, 11 | 1042 | |||
| AK172148.1 | - | 7, 8 | 1731 | |||
| BC120699.1 | - | 10, 11 | 1139 | 40129622 | ||
| BC137625.1 | - | 10, 11 | 1139 | 8860644 | ||

Clcn2
Genomic Map