siRNA Details
| Gene Symbol: Clcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009900.2 | NP_034030.2 | 22 | 3024 | ||
| GenBank mRNA | AF097415.1 | - | 22 | 2502 | ||
| AF139724.1 | - | 22 | 2453 | |||
| AK141656.1 | - | 23 | 3704 | |||
| AK155104.1 | - | 22 | 2318 | |||
| AK165000.1 | - | 20 | 2922 | |||
| AK171231.1 | - | 22 | 2318 | |||
| AK172148.1 | - | 18 | 3158 | |||
| BC120699.1 | - | 22 | 2415 | 40129622 | ||
| BC137625.1 | - | 22 | 2415 | 8860644 | ||
| CK633852.1 | - | 1 | 51 | |||

Clcn2
Genomic Map