siRNA Details
Gene Symbol: Clcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007711.3 | NP_031737.1 | 10 | 2366 | ||
NM_173873.1 | NP_776298.1 | 11 | 2542 | |||
NM_173874.1 | NP_776299.1 | 11 | 2542 | |||
NM_173876.3 | NP_776301.1 | 10 | 2087 | |||
GenBank mRNA | AF029347.1 | - | 10 | 1990 | ||
AK141136.1 | - | 11 | 2480 | |||
AK147364.1 | - | 9 | 6565 | |||
AK147548.1 | - | 9 | 6567 | |||
AK147621.1 | - | 9 | 6567 | |||
AK150688.1 | - | 11 | 2471 | |||
AK159698.1 | - | 11 | 2516 | |||
AK161327.1 | - | 11 | 2470 | |||
AK162639.1 | - | 11 | 2483 | |||
AK169297.1 | - | 11 | 2463 | |||
BC057133.1 | - | 10 | 2020 | 6833732 | ||
BC057855.1 | - | 10 | 2182 | |||
X78874.1 | - | 10 | 2191 |
