siRNA Details
| Gene Symbol: Clk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF033566.1 | - | 12 | 1328 | ||
| AK013974.2 | - | 10 | 1083 | |||
| AY653308.1 | - | 11 | 1218 | |||
| BC002220.1 | - | 8 | 884 | 3490616 | ||
| BC012675.1 | - | 12 | 1350 | 3982920 | ||
| CT010293.1 | - | 11 | 1250 | |||

Clk4
Genomic Map