siRNA Details
Gene Symbol: Clock | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289826.1 | NP_001276755.1 | 12 | 1185 | ||
NM_007715.6 | NP_031741.1 | 12 | 1228 | |||
GenBank mRNA | AB221635.1 | - | 9 | 840 | ||
AF000998.1 | - | 12 | 1228 | |||
AK030352.1 | - | 12 | 1128 | |||
AK129118.1 | - | 12 | 1185 | |||
BC129886.1 | - | 10 | 937 |
