siRNA Details
| Gene Symbol: Clock | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289826.1 | NP_001276755.1 | 20 | 2200 | ||
| NM_007715.6 | NP_031741.1 | 20 | 2243 | |||
| GenBank mRNA | AB221635.1 | - | 17 | 1855 | ||
| AF000998.1 | - | 20 | 2243 | |||
| AK129118.1 | - | 20 | 2200 | |||
| AY118227.1 | - | 5 | 605 | |||
| BC129886.1 | - | 18 | 1951 | |||

Clock
Genomic Map